missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BBS12 Polyclonal Antibody

Descripción
Recommended positive controls: HepG2. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The BBS12 gene is a key contributor to Bardet-Biedl Syndrome (BBS), a rare genetic disorder characterized by primary cilia dysfunction, leading to diverse clinical manifestations such as retinal degeneration, obesity, polydactyly, renal abnormalities, and learning difficulties. BBS12 is located on chromosome 4q27 and encodes a vertebrate-specific chaperonin-like protein. Together with BBS6 and BBS10, BBS12 forms a part of a complex associated with the CCT/TRiC family of chaperonins, which are essential for the proper assembly of the BBSome complex. This complex plays a critical role in the trafficking of vesicles to cilia and is crucial for ciliary function and structure. Mutations in BBS12 are responsible for approximately 5% of BBS cases, underscoring its significant role in the pathophysiology of the syndrome. These mutations often lead to disruptions in ciliary functions, which are pivotal for the syndrome's manifestation.
Especificaciones
Especificaciones
| Antígeno | BBS12 |
| Aplicaciones | Western Blot |
| Clasificación | Polyclonal |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 |
| génica | BBS12 |
| N.º de referencia del gen | Q6ZW61 |
| Alias de gen | Bardet-Biedl syndrome 12; Bardet-Biedl syndrome 12 (human); Bardet-Biedl syndrome 12 protein; Bardet-Biedl syndrome 12 protein homolog; Bbs12; C4orf24; DN-452A22.3; Gm1805; Gm407; Gm721 |
| Símbolos de los genes | BBS12 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?