Learn More
BAX Mouse anti-Human, CoraLite 488, Clone: 4G5E8, Proteintech
Código de producto. 16820616 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
16820616 100 μL 100 microlitros
1 options
Código de producto. 16820616
Proveedor: Proteintech CL48860267100UL
Mouse Monoclonal Antibody
The BAX antibody from Proteintech is a mouse monoclonal antibody generated with fusion protein of human BAX. This antibody recognizes human, mouse, rat antigen. The BAX antibody has been validated for the following applications: FC analysis. This antibody is coralite™488.
Especificaciones
Antígeno BAX
Aplicaciones Western Blot, Flow Cytometry
Clasificación Monoclonal
Clon 4G5E8
Conjugado CoraLite 488
Especie del huésped Mouse
Cantidad 100 μL
Primario o secundario Primary
ID de gen (Entrez) 581
Especies diana Human, Rat, Mouse
Isotype IgG2b