Learn More
Ansell Edmont™ Guantes de nitrilo desechables específicos para cada mano Barrier™
Manufactured from five layer film for wide spectrum chemical resistance. Ansell™ Barrier™ Disposable Hand Specific Nitrile Gloves can also be used as liners for more dangerous applications.
Marca:  Ansell Edmont™ 02-100_7
Código de producto. 11797373
Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
Ver productos alternativos »
Descripción
Descripción
  • Chemical resistant film gloves with an ergonomic design
  • Integral, non-woven liner absorbs and dissipates perspiration
  • Patented seam technology locks out chemicals and keeps the protective film and liner together as a single unit
  • Worn alone, Barrier™ gloves provide flexibility and tactility
  • Where the risk of physical damage is greater, the gloves should be worn as liners under heavier nitrile, neoprene, or PVC gloves when used for riskier applications where risk of phyiscal damage is greater