missing translation for 'onlineSavingsMsg'
Learn More

Balicatib, MedChemExpress

Código de producto. 16402437
Encompass_Preferred
Change view
Click to view available options
Cantidad:
5 mg
10 mg
50 mg
100 mg
Unit Size:
10 miligramos
100 miligramos
5 miligramos
50 miligramos
Product Code. Cantidad unitSize
16402437 50 mg 50 miligramos
16488897 5 mg 5 miligramos
16436636 100 mg 100 miligramos
16436318 10 mg 10 miligramos
4 options
This item is not returnable. View return policy

Product Code. 16402437

Brand: Medchem Express HY15100

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

MedChemExpress Balicatib (AAE581) is a potent, orally active and selective cathepsin K inhibitor with IC50 values of 22, 61, 48, 2900 nM for cathepsin K, cathepsin B, cathepsin L, cathepsin S, respectively. Balicatib inhibits bone turnover, decreases bone formation rates. Balicatib has the potential for the research of osteoporosis.

Balicatib (AAE581) is a potent, orally active and selective cathepsin K inhibitor with IC50 values of 22, 61, 48, 2900 nM for cathepsin K, cathepsin B, cathepsin L, cathepsin S, respectively. Balicatib inhibits bone turnover, decreases bone formation rates. Balicatib has the potential for the research of osteoporosis.

Specifications

Nombre del producto químico o material Balicatib
Forma física Solid
CAS 354813-19-7
Color Blanco
Cantidad 50 mg
Notas de grado de pureza Research
Fórmula molecular C23H33N5O2
Para utilizar con (aplicación) Neuroscience-Neuromodulation
Sinónimo AAE581
Información de solubilidad DMSO : 75 mg/mL (182.24 mM; Need ultrasonic)
SMILES CCCN1CCN(C2=CC=C(C(NC3(C(NCC#N)=O)CCCCC3)=O)C=C2)CC1
Peso molecular (g/mol) 411.54
Formula Weight (peso de la fórmula) 411.54
Porcentaje de pureza 99.07%
Grado Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.