Learn More
Invitrogen™ BAK1 Monoclonal Antibody (4C2)

Mouse Monoclonal Antibody
Marca: Invitrogen™ MA536225
Descripción
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human 293T whole cell, human A431 whole cell, human THP-1 whole cell, human MCF-7 whole cell, human PC-3 whole cell. IHC: human intestinal cancer tissue, human mammary cancer tissue, human tonsil tissue, human lung cancer tissue, mouse intestine tissue, rat intestine tissue. ICC/IF: A431 cell. Flow: PC-3 cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
The protein encoded by this gene belongs to the BCL2 protein family. BCL2 family members form oligomers or heterodimers and act as anti- or pro-apoptotic regulators that are involved in a wide variety of cellular activities. This protein localizes to mitochondria, and functions to induce apoptosis. It interacts with and accelerates the opening of the mitochondrial voltage-dependent anion channel, which leads to a loss in membrane potential and the release of cytochrome c. This protein also interacts with the tumor suppressor P53 after exposure to cell stress.
Especificaciones
| BAK1 | |
| Monoclonal | |
| 500 μg/mL | |
| PBS with 4mg trehalose and 0.05mg sodium azide | |
| O08734, Q16611 | |
| BAK1 | |
| E.coli-derived human BAK/BAK1 recombinant protein (Position: A22-S211). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG2b |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 4C2 | |
| Unconjugated | |
| BAK1 | |
| Apoptosis regulator BAK; BAK; Bak1; BAK-LIKE; BCL2 antagonist/killer 1; Bcl2 homologous antagonist/killer; Bcl-2 homologous antagonist/killer; BCL2-antagonist/killer 1; BCL2L7; bcl2-L-7; BCL2-like 7 protein; BCL2-like 7protein; bcl-2-like protein 7; Bcl-2-likeous antagonist/killer; CDN1; MGC117255; MGC3887; N-Bak; N-BAK1; pro-apoptotic protein BAK | |
| Mouse | |
| Antigen affinity chromatography | |
| RUO | |
| 116502, 12018, 578 | |
| -20°C | |
| Lyophilized |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.