Learn More
Invitrogen™ Bacteriophage M13 Recombinant Mouse Monoclonal Antibody (05), FITC, Invitrogen™
Descripción
Tail virion protein G7P (Bacteriophage M13) may initiate with G9P the virion concomitant assembly-budding process, by interacting with the packaging signal of the viral genome. The assembly-budding takes place at the host inner membrane. In turn, G7P and G9P are present at the end of the filamentous virion that emerges first from the bacterial host. Anti-Bacteriophage M13 Antibody is useful for researchers interested in HCP, Host Cell Membranes, Transmembrane Viruses, and E. coli bacteria.
Especificaciones
Especificaciones
| Antígeno | Bacteriophage M13 |
| Aplicaciones | Flow Cytometry |
| Clasificación | Recombinant Monoclonal |
| Clon | 05 |
| Concentración | 0.1 mg/mL |
| Conjugado | FITC |
| Formulación | PBS with no preservative |
| Alias de gen | Enterobacteria phage M13 |
| Especie del huésped | Mouse |
| Inmunógeno | M13 Bacteriophage. |
| Mostrar más |