missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ B7-H4 Recombinant Rabbit Monoclonal Antibody (BLR043F)
GREENER_CHOICE

Código de producto. 17775649
Change view
Click to view available options
Cantidad:
10 μL
100 μL
Tamaño de la unidad:
10 microlitros
100 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
17775649 100 μL 100 microlitros
17785649 10 μL 10 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 17775649 Proveedor Invitrogen™ N.º de proveedor MA544281

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Recombinant Monoclonal Antibody

Epitope retrieval with citrate buffer pH6.0 is recommended for FFPE cell sections.

B7-H4 (B7x, VTCN1) a costimulatory protein which is reported to function as a negative regulator of T-cell mediated immunity. Although B7-H4 binds an unknown receptor, it is thought to deliver an inhibitory signal to T-cells preventing their proliferation, cell cycle progression and interleukin-2 production. B7-H4 deficient mice are only minimally affected; suggesting B7-H4 is important in the fine tuning of the T-cell mediated immune response. B7-H4 is expressed on activated T-cells, B-cells, monocytes and dendritic cells. Aberrant expression has been associated with cancers of the lung, breast and ovary in humans. B7-H4 is expressed primarily on the membrane of lymphoid cells, and as an immunoinhibitory protein that interacts with receptors on the surface of T lymphocytes, it is involved in mediating cellular and humoral immune responses. Overexpression of B7-H4 is associated with certain malignancies, including ovarian and breast cancer, which may be a mechanism by which tumor cells suppress T cell immunity and facilitate tumor progression. Further, VTCN1 may be a useful biomarker for the early detection of ovarian cancer and Renal Cell Carcinoma (RCC).
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno B7-H4
Aplicaciones Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Clasificación Recombinant Monoclonal
Clon BLR043F
Concentración 50 μg/mL
Conjugado Unconjugated
Formulación BBS with 0.1% BSA and 0.09% sodium azide; pH 7.8-8.6
génica Vtcn1
N.º de referencia del gen Q7Z7D3
Alias de gen B7 family member, H4; B7 homolog 4; B7 superfamily member 1; B7h.5; B7H4; B7-H4; B7s1; B7x; BC032925; EMBL:AAH95842.1}; FLJ22418; immune costimulatory protein B7-H4; PRO1291; Protein B7S1; RGD1311204; RP11-229A19.4; t cell costimulatory molecule B7x; T-cell costimulatory molecule B7x; UNQ659/PRO1291; VCTN1; V-set domain containing T cell activation inhibitor 1; V-set domain containing T-cell activation inhibitor 1; V-set domain-containing T-cell activation inhibitor 1; Vtcn1; vtcn1 {ECO:0000312
Símbolos de los genes Vtcn1
Especie del huésped Rabbit
Inmunógeno Recombinant protein representing the extracellular domain of human B7-H4 (residues 30-282).
Método de purificación Purified
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 79679
Especies diana Human
Contenido y almacenamiento 4°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.