Learn More
B3GNT5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 20422-1-AP-150UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
B3GNT5 is a member of the beta-1, 3-N-acetylglucosaminyltransferase family. This enzyme is a type II membrane protein. It exhibits strong activity to transfer GlcNAc to glycolipid substrates and is identified as the most likely candidate for lactotriaosylceramide synthase. This enzyme is essential for the expression of Lewis X epitopes on glycolipids.Especificaciones
| B3GNT5 | |
| Polyclonal | |
| Unconjugated | |
| B3GNT5 | |
| B3GN T5, B3GNT5, Beta 1,3 Gn T5, beta3Gn T5, BGnT 5, Lactotriaosylceramide synthase, Lc(3)Cer synthase, Lc3 synthase | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 108105, 116740, 84002 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.19 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8BGY6, Q99NB2, Q9BYG0 | |
| B3GNT5 | |
| B3GNT5 Fusion Protein Ag14246 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |