Learn More
BUCHI FlashPure EcoFlex Cartridges, Silica Solid Loader
FlashPure cartridges are offered in a wide range of sizes, covering different stationary phases, particle sizes and geometries. This enables the user to choose the flash cartridge which best suits his purification needs.
Descripción
- Different sizes for scalability from milligrams to grams
- Broad range of different normal and reversed phases
- Small and spherical particles for fast separations and high loadings
- Compatibility with the Pure Navigator software (flash method optimization by interpreting TLC or HPLC scouting runs).
- RFID tag for an automatic transfer of important parameters for a run on a Pure chromatography system.
- Loading capacity: 0.1 - 10%
- Endcapped: No
Compliance
UL, CE, CSA