missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AZGP1 Monoclonal Antibody (21), Invitrogen™
Descripción
Specificity: Human Alpha-2-glycoprotein/AZGP1
Alpha-2-glycoprotein 1, zinc-binding (AZGP1) is a gene encoding a protein that belongs to the immunoglobulin superfamily. This protein is abundant in various body fluids, such as blood plasma and milk, and is involved in lipid mobilization. AZGP1 is expressed in numerous tissues, including the liver, prostate, and adipose tissue, where it plays a significant role in the regulation of lipid metabolism and adipocyte function. It acts as an adipokine, influencing the breakdown and mobilization of fats, thereby contributing to energy homeostasis. In cancer research, AZGP1 has been highlighted for its potential as a prognostic biomarker, particularly in prostate and breast cancer, where lower expression levels are often associated with poor prognosis. Recent studies have also linked AZGP1 to cachexia, an extreme form of weight loss and muscle wasting, by promoting lipid mobilization in adipose tissue and influencing systemic catabolic states. Due to its multifaceted roles in lipid metabolism and disease processes, AZGP1 is considered a promising target for therapeutic interventions in metabolic and cancer-related disorders.
Especificaciones
Especificaciones
| Antígeno | AZGP1 |
| Aplicaciones | ELISA |
| Clasificación | Monoclonal |
| Clon | 21 |
| Concentración | 1.59 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with no preservative |
| génica | AZGP1 |
| N.º de referencia del gen | P25311 |
| Alias de gen | alpha-2 - glycoprotein 1 zinc; alpha-2 - glycoprotein 1, zinc; alpha-2-glycoprotein 1, zinc; alpha-2-glycoprotein 1, zinc-binding; Alpha-2-glycoprotein, zinc; Azgp1; major histocompatibility complex class I-like protein; testicular tissue protein Li 227; ZA2G; ZA2GA; Zag; Zinc a 2 glycoprotein; Zinc a2glycoprotein; Zinc alpha2glycoprotein; zinc alpha-2-glycoprotein 1; Zinc a 2 glycoprotein; Zinc a2glycoprotein; zinc-alpha-2-glycoprotein; Zn - alpha2 - glycoprotein; Zn a 2 glycoprotein; Zn a 2 GP; Zn alpha 2 glycoprotein; Zn alpha 2 GP; Zn a 2 glycoprotein; Zn a 2 GP; Zna2gp; zn-alpha-2-glycoprotein; Zn-alpha2-glycoprotein; zn-alpha-2-GP; ZNGP1 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?