missing translation for 'onlineSavingsMsg'
Learn More

AZD-7648, MedChemExpress

Código de producto. 16464528
Encompass_Preferred
Click to view available options
Cantidad:
5 mg
10 mg
Tamaño de la unidad:
10 miligramos
5 miligramos
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 16464528

Marca: Medchem Express HY111783

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

MedChemExpress AZD-7648 is a potent, orally active, selective DNA-PK inhibitor with an IC50 of 0.6 nM. AZD-7648 induces apoptosis and shows antitumor activity.

AZD-7648 is a potent, orally active, selective DNA-PK inhibitor with an IC50 of 0.6 nM. AZD-7648 induces apoptosis and shows antitumor activity.

Specifications

Nombre del producto químico o material AZD-7648
CAS 2230820-11-6
Color Gris
Notas de grado de pureza Research
Fórmula molecular C18H20N8O2
Cantidad 10 mg
Información de solubilidad DMSO : 5 mg/mL (13.14 mM; ultrasonic and warming and heat to 80°C)
SMILES CC1=CC2=NC=NN2C=C1NC3=NC=C(N(C)C(N4C5CCOCC5)=O)C4=N3
Peso molecular (g/mol) 380.4
Formula Weight (peso de la fórmula) 380.4
Porcentaje de pureza 98.14%
Grado Research
Forma física Solid
Para utilizar con (aplicación) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.