missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AVEN Polyclonal Antibody

Descripción
Keep as concentrated solution. Positive Control: 293T, A431, HeLa, HepG2. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Apoptosis plays a major role in normal organism development, tissue homeostasis, and removal of damaged cells. Disruption of this process has been implicated in a variety of diseases such as cancer. Aven is a recently discovered protein that blocks apoptosis induced by Apaf-1 and caspase-9. It is thought that Aven functions by binding to Bcl-xL, an antiapoptotic member of the Bcl-2 family, and to Apaf-1, possibly interfering with the ability of Apaf-1 to self-associate, suggesting that Aven impedes Apaf-1-mediated caspase activation. Higher levels of Aven mRNA are seen in patients with acute leukemia than in control patients, suggesting that Aven may be useful as a prognostic indicator in leukemia patients.
Especificaciones
Especificaciones
| Antígeno | AVEN |
| Aplicaciones | Western Blot |
| Clasificación | Polyclonal |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 20% glycerol and 0.01% thimerosal; pH 7 |
| génica | AVEN |
| N.º de referencia del gen | Q9NQS1 |
| Alias de gen | 1700013A01Rik; 1700056A21Rik; apoptosis and caspase activation inhibitor; apoptosis, caspase activation inhibitor; AVEN; Cell death regulator Aven; mAven-L; mAven-S; PDCD12; programmed cell death 12 |
| Símbolos de los genes | AVEN |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?