missing translation for 'onlineSavingsMsg'
Learn More

Autotaxin modulator 1, MedChemExpress

Código de producto. 16455368
Encompass_Preferred
Change view
Click to view available options
Cantidad:
5 mg
10 mg
50 mg
Tamaño de la unidad:
10 miligramos
5 miligramos
50 miligramos
Product Code. Cantidad unitSize
16455368 10 mg 10 miligramos
16471557 50 mg 50 miligramos
16418087 5 mg 5 miligramos
3 options
This item is not returnable. View return policy

Product Code. 16455368

Brand: Medchem Express HY12812

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

MedChemExpress Autotaxin modulator 1 is an autotaxin (ATX) enzyme inhibitor, extracted from patent WO 2014018881 A1, Compound Example 12b. Autotaxin modulator 1 is expected to be useful for researching demyelination due to injury or disease, as well as for researching proliferative disorders such as cancer.

Autotaxin modulator 1 is an autotaxin (ATX) enzyme inhibitor, extracted from patent WO 2014018881 A1, Compound Example 12b. Autotaxin modulator 1 is expected to be useful for researching demyelination due to injury or disease, as well as for researching proliferative disorders such as cancer.

Specifications

Nombre del producto químico o material Autotaxin modulator 1
CAS 1548743-69-6
Color Blanco
Notas de grado de pureza Research
Fórmula molecular C28H31F6NO3
Cantidad 10 mg
Información de solubilidad DMSO : 100 mg/mL (183.98 mM; Need ultrasonic)
SMILES O=C(C1CC(N2[C@@H](C3=CC=C4C=CC(O[C@H]5CC[C@@H](C(F)(F)F)CC5)=C(C(F)(F)F)C4=C3)C)CCC2C1)O
Peso molecular (g/mol) 543.54
Formula Weight (peso de la fórmula) 543.54
Porcentaje de pureza 98.93%
Grado Research
Forma física Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.