missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ATP6V0D1 Polyclonal Antibody
GREENER_CHOICE

Código de producto. 15556972
Change view
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
Código de producto. Cantidad unitSize
15556972 100 μL 100 microlitros
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 15556972

Marca: Invitrogen™ PA545852

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

Peptide sequence: KLLFEGAGSN PGDKTLEDRF FEHEVKLNKL AFLNQFHFGV FYAFVKLKEQ Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.

This gene encodes a component of vacuolar ATPase (V-ATPase), a multisubunit enzyme that mediates acidification of eukaryotic intracellular organelles. V-ATPase dependent organelle acidification is necessary for such intracellular processes as protein sorting, zymogen activation, receptor-mediated endocytosis, and synaptic vesicle proton gradient generation. V-ATPase is composed of a cytosolic V1 domain and a transmembrane V0 domain. The V1 domain consists of three A and three B subunits, two G subunits plus the C, D, E, F, and H subunits. The V1 domain contains the ATP catalytic site. The V0 domain consists of five different subunits: a, c, c', and d. Additional isoforms of many of the V1 and V0 subunit proteins are encoded by multiple genes or alternatively spliced transcript variants. This encoded protein is known as the D subunit and is found ubiquitously.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno ATP6V0D1
Aplicaciones Western Blot
Clasificación Polyclonal
Concentración 0.5 mg/mL
Conjugado Unconjugated
Formulación PBS with 2% sucrose and 0.09% sodium azide
génica ATP6V0D1
N.º de referencia del gen P51863, P61421
Alias de gen 32 kd accessory protein; 32 kDa accessory protein; Ac39; AI267038; Atp6d; ATP6DV; Atp6v0d1; ATPase H+ transporting V0 subunit d1; ATPase, H+ transporting, lysosomal (vacuolar proton pump), 42 kDa; ATPase, H+ transporting, lysosomal (vacuolar proton pump), member D; ATPase, H+ transporting, lysosomal 38kDa, V0 subunit D; ATPase, H+ transporting, lysosomal 38kDa, V0 subunit d1; ATPase, H+ transporting, lysosomal V0 subunit D1; ATPase, H+ transporting, lysosomal, V0 subunit d1; ATPase, H+ transporting, V0 subunit D; cto; fb73h07; FLJ43534; H(+)-transporting two-sector ATPase, subunit D; lysosomal 38kDa; P39; Physophilin; Unknown (protein for MGC:63769); Vacuolar proton pump subunit d 1; vacuolar proton-ATPase, subunit D; V-ATPase, subunit D; V-ATPase 40 KDa accessory protein; V-ATPase AC39 subunit; V-ATPase subunit d 1; V-ATPase, subunit D; VATX; Vma6; VPATPD; V-type proton ATPa; V-type proton ATPase subunit d 1; wu:fb73h07; zgc:63769
Símbolos de los genes ATP6V0D1
Especie del huésped Rabbit
Inmunógeno Synthetic peptide directed towards the C-terminal of human ATP6V0D1 (aa 275-324).
Método de purificación Affinity chromatography
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 11972, 9114
Especies diana Human, Mouse
Contenido y almacenamiento -20°C, Avoid Freeze/Thaw Cycles
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.