missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
Positive Samples: Mouse brain, Mouse heart, Mouse kidney, Mouse liver, Rat liver Immunogen sequence: SGTGIAAMSV MRPE.
ATP6V0C is a component of vacuolar ATPase (V-ATPase), a multisubunit enzyme that mediates acidification of eukaryotic intracellular organelles. V-ATPase dependent organelle acidification is necessary for such intracellular processes as protein sorting, zymogen activation, receptor-mediated endocytosis, and synaptic vesicle proton gradient generation. V-ATPase is composed of a cytosolic V1 domain and a transmembrane V0 domain. The V1 domain consists of three A and three B subunits, two G subunits plus the C, D, E, F, and H subunits. The V1 domain contains the ATP catalytic site. The V0 domain consists of five different subunits: a, c, c', c', and d. ATP6V0C encodes the V0 subunit c.
Especificaciones
Especificaciones
| Antígeno | ATP6V0C |
| Aplicaciones | ELISA, Western Blot, Immunocytochemistry |
| Clasificación | Polyclonal |
| Concentración | 1.37 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 |
| génica | ATP6V0C |
| N.º de referencia del gen | P27449, P63081, P63082 |
| Alias de gen | Atp6c; Atp6c2; Atp6l; ATP6V0C; ATPase H+ transporting V0 subunit c; ATPase, H+ transporting, lysosomal (vacuolar proton pump) 16 kDa; ATPase, H+ transporting, lysosomal (vacuolar proton pump), subunit C; ATPase, H+ transporting, lysosomal 1; ATPase, H+ transporting, lysosomal 16kD, V0 subunit C; ATPase, H+ transporting, lysosomal 16kDa, V0 subunit c; ATPase, H+ transporting, lysosomal 2; ATPase, H+ transporting, lysosomal V0 subunit C; ATPase, H+ transporting, V0 subunit C; ATPL; Atpl-rs1; H(+)-ATPase (mvp); H(+)-transporting two-sector ATPase, 16 kDa subunit; Mvp; PL16; vacuolar ATP synthase 16 kDa proteolipid subunit; vacuolar H+ ATPase proton channel subunit; vacuolar proton pump 16 kDa proteolipid subunit; VATL; V-ATPase 16 kDa proteolipid subunit; Vma3; VPPC; V-type proton ATPase 16 kDa proteolipid subunit |
| Símbolos de los genes | ATP6V0C |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?
