missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATP5C1 Mouse anti-Human, Clone: 1B5A2, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 60284-1-IG-150UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
This gene encodes a subunit of mitochondrial ATP synthase. Mitochondrial ATP synthase catalyzes ATP synthesis, utilizing an electrochemical gradient of protons across the inner membrane during oxidative phosphorylation. ATP synthase is composed of two linked multi-subunit complexes: the soluble catalytic core, F1, and the membrane-spanning component, Fo, comprising the proton channel. The catalytic portion of mitochondrial ATP synthase consists of 5 different subunits (alpha, beta, gamma, delta, and epsilon) assembled with a stoichiometry of 3 alpha, 3 beta, and a single representative of the other 3. The proton channel consists of three main subunits (a, b, c). This gene encodes the gamma subunit of the catalytic core. Alternatively spliced transcript variants encoding different isoforms have been identified. This gene also has a pseudogene on chromosome 14.Especificaciones
| ATP5C1 | |
| Monoclonal | |
| 0.77 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P36542 | |
| ATP5C1 | |
| ATP5C1 Fusion Protein Ag1343 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1B5A2 | |
| Unconjugated | |
| ATP5C1 | |
| ATP5C, ATP5C1, ATP5CL1, F ATPase gamma subunit | |
| Mouse | |
| Protein A | |
| RUO | |
| 509 | |
| -20°C | |
| Liquid |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido