Learn More
ATP2A1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Código de producto. 16871895 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
16871895 20 μL 20 microlitros
16861895 150 μL 150 microlitros
2 options
Código de producto. 16871895
Marca: Proteintech 223611AP20UL
Rabbit Polyclonal Antibody
ATP dependent calcium pumps are responsible, in part, for the maintece of low cytoplasmic free calcium concentrations. The ATP pumps that reside in intracellular organelles are encoded by a family of structurally related enzymes, termed the sarcoplasmic or endoplasmic reticulum calcium (SERCA) ATPases. The SERCA1 gene is exclusively expressed in type II (fast) skeletal muscle. The SERCA2 gene is subject to tissue dependent processing which is responsible for the generation of SERCA2a muscle-specific form expressed in type I (slow) skeletal, cardiac and smooth muscle and the SERCA2b isoform expressed in all cell types. The SERCA3 gene is not as well characterized and is found in non-muscle cells.
Especificaciones
Antígeno ATP2A1
Aplicaciones Immunohistochemistry (Paraffin), Immunoprecipitation
Clasificación Polyclonal
Concentración 0.38 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
génica ATP2A1
N.º de referencia del gen O14983, Q8R429
Alias de gen ATP2A, ATP2A1, Calcium pump 1, SERCA1, SR Ca(2+) ATPase 1
Símbolos de los genes ATP2A1
Especie del huésped Rabbit
Inmunógeno ATP2A1 Fusion Protein Ag17944
Método de purificación Antigen Affinity Chromatography
Cantidad 20 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 11937, 487
Especies diana Human, Mouse
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG