missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ATG2B Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA534428
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
A suggested positive control is K562 cell lysate. PA5-34428 can be used with blocking peptide PEP-1470.
Autophagy, the process of bulk degradation of cellular proteins through an autophagosomic-lysosomal pathway is important for normal growth control and may be defective in tumor cells. It is involved in the preservation of cellular nutrients under starvation conditions as well as the normal turnover of cytosolic components. This process is negatively regulated by TOR (Target of rapamycin) through phosphorylation of autophagy protein APG1. Another member of the autophagy family of proteins is ATG2B, one of two homologs of ATG2 that is essential for autophagosome formation and important for regulation of size and distribution of lipid droplets. Relatively high rates of ATG2B mutations were observed in gastric and colorectal carcinomas, suggesting that deregulating the autophagy process may contribute to cancer development.
Especificaciones
| ATG2B | |
| Polyclonal | |
| Unconjugated | |
| ATG2B | |
| 2410024A21Rik; AI047755; AI503411; ATG2 autophagy related 2 homolog B; Atg2b; autophagy related 2B; autophagy-related protein 2 homolog B; AW558123; C030004M05Rik; C14orf103; C630028L02Rik; mKIAA4067; RGD1304878 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 55102 | |
| Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q96BY7 | |
| ATG2B | |
| A 19 amino acid synthetic peptide near the amino terminus of human ATG2B. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido