missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ATG14 Polyclonal Antibody

Descripción
Keep as concentrated solution. Predicted reactivity: Mouse (93%), Rat (91%), Bovine (94%). Positive Control: Huh7, HeLa. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
ATG14 (Autophagy related protein homolog 14) is encoded by the ATG14 gene and is located on chromosome 14 in humans. It is also known as ATG14L or BARKOR (Beclin 1-associated autophagy-related key regulator). Autophagy is an evolutionarily conserved process that involves recycling of misfolded cellular proteins and degradation of dysfunctional organelles. Autophagy is induced as a cellular response to nutrient stress which includes the formation of autophagosomes, fusion of these with the lysosome and formation of the autophagolysosome. The Class III PI3-Kinase, Vps34 and its interacting partner Beclin 1 have been shown to form a complex which specifically includes ATG 14 (type 1) or UVRAG (type 2). Atg 14 guides the type 1 complex to the preautophagosomal structure (PAS) and is critical for the initiation of autophagosome formation.
Especificaciones
Especificaciones
| Antígeno | ATG14 |
| Aplicaciones | Western Blot |
| Clasificación | Polyclonal |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 |
| génica | Atg14 |
| N.º de referencia del gen | Q6ZNE5 |
| Alias de gen | 4832427M01; Atg14; ATG14 autophagy related 14 homolog; Atg14L; Atg14-like protein; autophagy related 14; autophagy-related protein 14-like protein; Bakor; Barkor; beclin 1-associated autophagy-related key regulator; Beclin 1-Interacting protein; D14Ertd114e; D14Ertd436e; Kiaa0831; RGD1304610; VATG14 autophagy related 14 homolog |
| Símbolos de los genes | Atg14 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?