Learn More
CPC (Colder) AseptiQuik™ STC II Genderless Quick-Disconnect Fittings, AspetiQuik™ S to Sanitary Adapter, Polycarbonate
Código de producto. 30053179
Change view
Click to view available options
Diámetro interior (imperial):
1-1/2 in
3/4 in
Tamaño de la unidad:
cada uno
Código de producto. Diámetro interior (imperial) unitSize
30053179 3/4 in cada uno
30053916 1-1/2 in cada uno
2 options
Código de producto. 30053179
Marca: CPC (Colder) 5011202
Solo quedan null
Añadir a la cesta
Añadir a la cesta
Low-flow genderless sterile connector and Steam-Thru™ II connector in a single unit simplify process integration while maximizing flexibility
Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application. Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants. Simple flip, click, and pull design minimizes operator errors. Create quick and easy sterile connections, even in nonsterile environments. Robust construction provides reliable and repeatable fluid transfer. Specifically designed for small-volume fluid transfer applications. Innovative three-port steam design. Sterilize AseptikQuik STC II fitting on stainless steel equipment via SIP. 0.375 in. nominal flow path size
Especificaciones
Certificaciones/conformidad ADCF;FDA;Non-DEHP;Non-Phthalate;RoHS;USP
Diámetro interior (imperial) 3/4 in
Temperatura mín. de funcionamiento (métrico) 1°C
Cantidad de embalaje 1 Ea.
Puerto de salida Quick Disconnect x Sanitary x Sanitary
Método de esterilización 130°C
Forma Straight
Incluye 3/4 in. Sanitary Clamp Size
Material (sello) Platinum-cured Silicone
Color White with blue pull tab and amber tint