Learn More
CPC (Colder) AseptiQuik™ S Low-Flow Genderless Quick-Disconnect Fittings, Hose Barb Adapter, Polycarbonate
Descripción
Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application. Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants. Simple flip, click, and pull design minimizes operator errors. Create quick and easy sterile connections, even in nonsterile environments. Specifically designed for small-volume fluid transfer applications. Robust construction provides reliable and repeatable fluid transfer. 0.25 in. nominal flow path size
Especificaciones
Especificaciones
| Certificaciones/conformidad | ADCF;FDA;Non-DEHP;Non-Phthalate;RoHS;USP |
| Diámetro interior (imperial) | 3/8 in |
| Cantidad de embalaje | 1 Ea. |
| Puerto de salida | Quick Disconnect x Hose Barb |
| Método de esterilización | Gamma irradiation up to 50 kGy |
| Forma | Straight |
| Material (sello) | Platinum-cured Silicone |
| Color | White with blue pull tab |