Learn More
CPC (Colder) AseptiQuik™ G Genderless Quick-Disconnect Fittings, Sanitary Adapter, Polycarbonate
Click to view available options
Incluye:
1-1/2 in Tamaño de la abrazadera sanitaria
3/4 in Tamaño de la abrazadera sanitaria
Tamaño de la unidad:
cada uno
Descripción
Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application. Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants. Simple flip, click, and pull design minimizes operator errors. Create quick and easy sterile connections, even in nonsterile environments. Robust construction provides reliable and repeatable fluid transfer. 0.5 in. nominal flow path size
Especificaciones
Especificaciones
| Certificaciones/conformidad | ADCF;FDA;Non-DEHP;Non-Phthalate;RoHS;USP |
| Temperatura mín. de funcionamiento (métrico) | 104°C |
| Cantidad de embalaje | 1 Ea. |
| Puerto de salida | Quick Disconnect x Sanitary |
| Método de esterilización | Gamma irradiation up to 50 kGy |
| Forma | Straight |
| Incluye | 3/4 in Tamaño de la abrazadera sanitaria |
| Material (sello) | Platinum-cured Silicone |
| Color | White with blue pull tab |
| Presión máxima | 60 psi. |
| Mostrar más |