missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ASCT2 Recombinant Rabbit Monoclonal Antibody (HL1988)

Descripción
Store as concentrated solution. Centrifuge briefly prior to opening vial.
SLC1A5 (Solute carrier family 1 (neutral amino acid transporter), member 5) is a member of the Na(+)-dependent amino acid transporter superfamily. It has a broad substrate specificity, a preference for zwitterionic amino acids, and a sodium-dependence. It accepts as substrates all neutral amino acids, including glutamine, asparagine, and branched-chain and aromatic amino acids, and excludes methylated amino acids, anionic amino acids, and cationic amino acids. It acts as a cell surface receptor for feline endogenous virus RD114, baboon M7 endogenous virus and type D simian retroviruses.
Especificaciones
Especificaciones
| Antígeno | ASCT2 |
| Aplicaciones | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Clasificación | Recombinant Monoclonal |
| Clon | HL1988 |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with no preservative |
| génica | SLC1A5 |
| N.º de referencia del gen | Q15758 |
| Alias de gen | AAAT; ASC-like Na(+)-dependent neutral amino acid transporter ASCT2; Asct2; ATB(0); ATB0; ATBO; baboon M7 virus receptor; CAZ-associated structural protein; H4-ASCT2; H4-system ASC-like transporter; insulin-activated amino acid transporter; M7V1; M7VS1; neutral amino acid transporter B; neutral amino acid transporter B(0); R16; RD114 virus receptor; RD114/simian type D retrovirus receptor; RDR; RDRC; SLC1A5; Slc1a7; sodium-dependent neutral amino acid transporter ASCT2; sodium-dependent neutral amino acid transporter type 2; solute carrier family 1 (neutral amino acid transporter), member 5; solute carrier family 1 member 5; solute carrier family 1, member 7 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?