Learn More
ASCC1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 12301-1-AP-150UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
This gene encodes a subunit of the activating signal cointegrator 1 (ASC-1) complex. The ASC-1 complex is a transcriptional coactivator that plays an important role in gene transactivation by multiple transcription factors including activating protein 1 (AP-1), nuclear factor kappa-B (NF-kB) and serum response factor (SRF). The encoded protein contains an N-terminal KH-type RNA-binding motif which is required for AP-1 transactivation by the ASC-1 complex. Mutations in this gene are associated with Barrett esophagus and esophageal adenocarcinoma. Alternatively spliced transcripts encoding multiple isoforms have been observed for this gene.Especificaciones
| ASCC1 | |
| Polyclonal | |
| Unconjugated | |
| ASCC1 | |
| ASC 1 complex subunit p50, ASC1p50, ASCC1, CGI 18, RP11 150D20.4, Trip4 complex subunit p50 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 294512, 51008, 69090 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8N9N2, Q9D8Z1 | |
| ASCC1 | |
| ASCC1 Fusion Protein Ag2946 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |