missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ASAHL Polyclonal Antibody
Descripción
In Western blots, approximately 25% cross-reactivity with recombinant mouse ASAHL is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
N-acylsphingosine amidohydrolase-like 1 (ASAHL), commonly referred to as acid ceramidase-like protein, is a gene located on human chromosome 22q13.31. ASAHL encodes a protein that functions as a ceramidase, responsible for hydrolyzing ceramide into sphingosine and fatty acid, which are pivotal for cell signaling pathways that regulate cell growth, differentiation, and apoptosis. Though similar to acid ceramidase, ASAHL's specific biological role and tissue distribution remain active research areas, suggesting potential involvement in lipid metabolism and cellular homeostasis. Dysregulation of ceramidase activity, including ASAHL, is linked to various diseases, such as cancer, neurodegenerative disorders, and metabolic syndromes, due to the crucial role of sphingolipid signaling in these conditions. The gene's activity may influence ceramide levels within cells, impacting susceptibility to apoptosis and other cell fate decisions, highlighting its importance in pathophysiological contexts.
Especificaciones
Especificaciones
| Antígeno | ASAHL |
| Aplicaciones | Immunoprecipitation, Western Blot |
| Clasificación | Polyclonal |
| Concentración | 0.2 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 5% trehalose and No Preservative |
| génica | Naaa |
| N.º de referencia del gen | Q02083 |
| Alias de gen | 2210023K21Rik; 3830414F09Rik; acid ceramidase-like protein; Asahl; ASAH-like protein; LOW QUALITY PROTEIN: N-acylethanolamine-hydrolyzing acid amidase; Naaa; N-acylethanolamine acid amidase; N-acylethanolamine-hydrolyzing acid amidase; N-acylethanolamine-hydrolyzing acid amidase subunit alpha; N-acylethanolamine-hydrolyzing acid amidase subunit beta; N-acylsphingosine amidohydrolase-like; PLT |
| Símbolos de los genes | Naaa |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?