Learn More
ARL3 Antibody, DyLight 650, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca: Novus Biologicals NBP2-97379C
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
ARL3 Polyclonal antibody specifically detects ARL3 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Especificaciones
| ARL3 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| DyLight 650 | |
| ADP-ribosylation factor-like 3, ARFL3ADP-ribosylation factor-like protein 3, ARF-like 3 | |
| Produced in rabbits immunized with E. coli-derived Human ARL3 fragment. | |
| 0.1 mL | |
| Signal Transduction | |
| 403 | |
| Store at 4°C in the dark. | |
| IgG |