Learn More
STEMCELL Technologies ArciTect™ T7 Endonuclease I Kit
Código de producto. 17129311
Encompass_Preferred
Change view
Click to view available options
Cantidad:
125 reacciones
25 reacciones
Tamaño de la unidad:
1 set
17129311 25 reacciones 1 set
17139311 125 reacciones 1 set
2 options
Código de producto. 17129311
Marca: STEMCELL Technologies 76021
For estimation of CRISPR-Cas9 genome editing efficiency
  • ArciTect™ T7 Endonuclease I is the preferred enzyme for detecting genome editing such as insertions or deletions (INDELs) generated by CRISPR-Cas9
  • ArciTect™ T7 Endonuclease I Kit is comprised of ArciTect™ T7 Endonuclease I and ArciTect™ T7 Endonuclease I Buffer (10X), which have been tested and validated for use with the ArciTect™ CRISPR-Cas9 genome editing system
  • ArciTect™ T7 Endonuclease I recognizes and cleaves mismatched DNA, cruciform DNA structures, Holliday structures or junctions, heteroduplex DNA, and, less efficiently, nicked double-stranded DNA
  • Since the cleavage efficiency is proportional to the number of INDELs created at a specific DNA target, ArciTect™ T7 Endonuclease I Kit is used to estimate gene-editing efficiency in a rapid and cost-effective manner
Especificaciones
Para utilizar con (aplicación) Estimation of CRISPR-Cas9 Genome Editing Efficiency
Cantidad 25 reacciones