Learn More
Chemsplash AquaRepel Non Toxic, Water Repellant Coverall
Descripción
Chemsplash Aqua Repel Coverall
- 40 GSM Polypropylene Coated Fabric
- Non Toxic, Waterproof Fabric
- Hood With Elasticated Band
- Elasticated Cuffs and Ankles
- Half Elasticated Waist
- Zipped Front With Adhesive Zip Flap
- PFAS Free
Description: The Chemsplash Aqua Repel Coverall is made from 40 GSM Polypropylene Coated Fabric. This is a Cat 1 suit which is lightweight and water repellent, providing protection against non-hazardous limited liquid splash.
Especificaciones
Especificaciones
| Tipo de producto | Hooded Coveralls |
| Tamaño | M |
| Sexo | Unisex |
| Material | Polipropileno |
| Color | Azul |
| Desechables | Single-use |
| Estilo de cintura | Elástico |
| Estilo de tobillo | Elastic |
| Estilo de manguito | Elastic |
| Estilo de cuello | Hooded |
| Mostrar más |