missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aquaporin 4 Polyclonal Antibody, Invitrogen™
Descripción
Product is shipped at room temperature as a lyophilized powder and should be stored at -20 °C upon receipt. Reconstitution: add 50 μL of deionized water.
In skeletal muscle, AQP4 (aquaporin 4 also known as mercurial insensitive water channel), localizes to the sarcolemma of fast-twitch muscle fibers. Aquaporins (AQPs) are a large family of integral membrane water transport channel proteins that facilitate the transport of water through the cell membrane. This function is conserved in animals, plants and bacteria. Many isoforms of aquaporin have been identified in mammals, designated AQP0 through AQP10. Aquaporins are widely distributed and it is not uncommon for more than one type of AQP to be present in the same cell. Although most aquaporins are only permeable to water, AQP3, AQP7, AQP9 and one of the two AQP10 transcripts are also permeable to urea and glycerol.
Especificaciones
Especificaciones
| Antígeno | Aquaporin 4 |
| Aplicaciones | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Western Blot |
| Clasificación | Polyclonal |
| Concentración | 0.8 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 1% BSA and 0.05% sodium azide; pH 7.4 |
| génica | AQP4 |
| N.º de referencia del gen | P47863, P55087, P55088 |
| Alias de gen | Aquaporin 4 |
| Símbolos de los genes | AQP4 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?