missing translation for 'onlineSavingsMsg'
Learn More

AQP7 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Product Code. 16843675 Shop All Proteintech Products
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Unit Size:
150 microlitros
20 microlitros
Product Code. Cantidad unitSize
16843675 20 μL 20 microlitros
16833675 150 μL 150 microlitros
2 options
This item is not returnable. View return policy

Product Code. 16843675

Brand: Proteintech 251311AP20UL

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Aquaporins/major intrinsic protein (MIP) are a family of water-selective membrane channels. Aquaporin 7 has greater sequence similarity with AQP3 and AQP9 and they may be a subfamily. Aquaporin 7 and AQP3 are at the same chromosomal location suggesting that 9p13 may be a site of an aquaporin cluster. Aquaporin 7 facilitates water, glycerol and urea transport. It may play an important role in sperm function.

Specifications

Antígeno AQP7
Aplicaciones Immunohistochemistry (Paraffin), Immunoprecipitation
Clasificación Polyclonal
Concentración 0.17 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
génica AQP7
N.º de referencia del gen O14520, O54794
Alias de gen AQP 7, AQP7, AQP7L, AQP9, AQPap, Aquaglyceroporin 7, aquaporin 7, Aquaporin 7 like, Aquaporin adipose
Símbolos de los genes AQP7
Especie del huésped Rabbit
Inmunógeno AQP7 Fusion Protein Ag17945
Método de purificación Antigen Affinity Chromatography
Cantidad 20 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 11832, 364
Especies diana Mouse, Human
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.