Learn More
APXL Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Marca: Novus Biologicals NB100-1350
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
APXL Polyclonal antibody specifically detects APXL in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide ELISA
Especificaciones
| APXL | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA | |
| apical protein, Xenopus laevis-like, apical protein-like (Xenopus laevis), Apical-like protein, APX homolog of Xenopus, APXLHSAPXL, DKFZp781J074, FLJ39277, Protein APXL, protein Shroom2, shroom family member 2 | |
| Peptide with sequence KCLLDSLQPERGK corresponding to C-Terminus according to NP_001640. | |
| 0.1 mg | |
| Vision | |
| 357 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide ELISA | |
| 0.5 mg/mL | |
| Immunohistochemistry 4 - 6 μg/mL, Immunohistochemistry-Paraffin 1:10-1:500, Peptide ELISA Detection limit 1:32000 | |
| NP_001640. | |
| Goat | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |