Learn More
Tocris Bioscience™ Aprotinin
GREENER_CHOICE
Competitive serine protease inhibitor
Marca:  Tocris Bioscience™ 4139/10
Encompass
Product Code. 15786545
Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
View alternative products »
This item is not returnable. View return policy
Description
Description
Aprotinin is a competitive serine protease inhibitor. Reversibly binds to and blocks the enzymatic active site. Inhibits a range of serine proteases including trypsin, chymotrypsin, kallikrein and plasmin. Inhibits cytopathogenic effect of SARS-CoV-2 and double-stranded RNA formation in SARS-CoV-2-infected cells.
This product is from Bovine Lung.
Specifications
Specifications
10 mg
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions