missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APP 695+1 Mouse anti-Human, Clone: 128715, R&D Systems™

Mouse Monoclonal Antibody
Marca: R&D Systems MAB890-SP
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
APP 695+1 Monoclonal specifically detects APP 695+1 in Human samples. It is validated for Western Blot, Immunoprecipitation.Especificaciones
| APP 695+1 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| APP | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human APP695 +1 frameshift mutant Leu18-Arg336 (predicted) Accession # NP_958817 | |
| 25 μg | |
| Primary | |
| Detects human APP695 +1 in Western blots. It recognizes a common epitope present on APP695 +1 and APP770 found within the segments L18-R288 or P365-R411 of APP770 (Accession # P05067). | |
| Human | |
| Purified |
| Western Blot, Immunoprecipitation | |
| 128715 | |
| Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
| NP_958817 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 351 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?