Learn More
APOOL Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 28514-1-AP-150UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
The APOOL antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human APOOL. This antibody recognizes human, mouse, rat antigen. The APOOL antibody has been validated for the following applications: WB, IHC, IF/ICC, ELISA analysis.Especificaciones
| APOOL | |
| Polyclonal | |
| Unconjugated | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 139322 | |
| Store at -20°C. Stable for one year after shipment. | |
| Polyclonal Antibody | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, ELISA | |
| 300 μg/mL | |
| CXorf33; FAM121A; MGC129748; UNQ8193 | |
| Recombinant Protein | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| 30 kDa | |
| Liquid |