Learn More
Invitrogen™ Apolipoprotein C2 Human ELISA Kit
Sandwich ELISA Kit
Marca:  Invitrogen™ EHAPOC2X10
Detalles adicionales : Peso : 2.50000kg
 Ver más versiones de este producto
Código de producto. 17834180
  • 4790.00€ / paquete de 10
Fecha estimada de envío: 05-05-2023
para ver el stock
Añadir a la cesta
Descripción
Descripción
The Human Apolipoprotein C2 (Hu ApoC2) ELISA quantitates Hu ApoC2 in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu ApoC2. Principle of the method The Human ApoC2 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Apolipoproteins are protein components of plasma lipoproteins. The apolipoprotein C gene family encodes four homologous proteins designated apoC-I to -IV, which specifically modulate the metabolism of triglyceride-rich lipoproteins. The human apoC-I gene maps to chromosome 19q13.2 and is expressed primarily in the liver where it is activated when monocytes differentiate into macrophages. The human apoC-II gene maps to chromosome 19q13.2 and encodes a 79 amino acid single chain protein that is a necessary cofactor for the activation of lipoprotein lipase, the enzyme that hydrolyzes triglycerides in plasma and transfers the fatty acids to tissues .
Especificaciones
Especificaciones
P02655
1.5 ng/mL
Solid-phase sandwich ELISA
ELISA Kit
Supernatant
10 x 96 assays
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis
344
<12%
Assay Diluent concentrate, Adhesive Plate Covers, Stop Solution, Standard, SAV-HRP, Pre-coated 96 well plate, Biotinylated Detection Antibody, Chromogen, Wash Buffer
10 x 96 Tests
Cardiovascular Research
2°C to 8°C
Human
4 hr. 45 min.
1.56 to 100 ng/mL
1.5 ng/mL to 100 ng/mL
Biotin
Other Proteins
This ELISA kit shows no cross-reactivity with the following cytokines tested: human Alpha-2 Microglobulin, ApoA, ApoC1, ApoC3, ApoE, ApoH, Artemin, BMP-6, Calbindin, CK beta 8-1 (CCL23), CNTF, EMMPRIN (CD147), Endostatin, Fibrinogen, FLRG, TNFSF18 (GITR Ligand), GPBB, Hemoglobin, HTRA2, IL-17 E, IL-1 R3 (IL1 R Acp), Lactoferrin, Mesothelin, PAPP-A, PD-ECGF (Thymidine Phosphorylase), PDGF R alpha, Pepsinogen 2, Periostin, Persephin, PTH, RAP, SAA (Serum Amyloid A), Serpin F1 (PEDF), Tie-1, TPA, Thrombospondin 2, Ubiquitin+1, Vitronectin, VWF
Human
Colorimetric Microplate Reader
APO-CII,APOC-II
<10%
HRP
RUO
Plasma, 2 μL; Serum, 2 μL; Supernatant, 100 μL
APO-CII, APOC-II
1 hr. 20 min.