Learn More
Apolipoprotein AI Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 14427-1-AP-20UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
This antibody was generated against the C-terminal region of human ApoA1.
Apolipoprotein A-1, also known as APOA1, is a human protein with a specific role in lipid metabolism. It binds to lipopolysaccharide or endotoxin, and has a major role in the anti-endotoxin function of HDL. The gene is mapped to 11q23. And it is a single polypeptide chain with 243 amino acid residues of known primary amino acid sequence. The ApoA-I protein promotes cholesterol efflux from tissues to the liver for excretion. It is a cofactor for lecithin cholesterolacyltransferase (LCAT) which is responsible for the formation of most plasma cholesteryl esters. ApoA-I is also isolated as a prostacyclin (PGI2) stabilizing factor, and thus may have an anticlotting effect. Defects in the gene encoding it are associated with HDL deficiencies, including Tangier disease, and with systemic non-neuropathic amyloidosis. Additionally, ApoA-I overexpression promotes macrophage-specific reverse cholesterol transport.Especificaciones
| Apolipoprotein AI | |
| Polyclonal | |
| Unconjugated | |
| APOA1 | |
| Apo AI, ApoA I, APOA1, apolipoprotein A I, Apolipoprotein A1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11806, 335 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry, Immunoprecipitation, Immunofluorescence, Flow Cytometry | |
| 0.52 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P02647, Q00623 | |
| Apoa1 | |
| Apolipoprotein AI Fusion Protein Ag5793 | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |