missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ APOBEC3A Polyclonal Antibody

Descripción
APOBEC3A Polyclonal Antibody for Western Blot, IHC (P)
PHO1 a member of the cytidine deaminase gene family. The PHO1 gene is one of seven related genes or pseudogenes found in a cluster, thought to result from gene duplication, on chromosome 22. Members of the cluster encode proteins that are structurally and functionally related to the C to U RNA-editing cytidine deaminase APOBEC1. It is thought that the proteins may be RNA editing enzymes and have roles in growth or cell cycle control. This gene encodes a protein that lacks the zinc binding activity and may be an expressed pseudogene.
Especificaciones
Especificaciones
| Antígeno | APOBEC3A |
| Aplicaciones | Immunohistochemistry (Paraffin), Western Blot |
| Clasificación | Polyclonal |
| Concentración | 2 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 0.09% sodium azide; pH 7.4 |
| génica | APOBEC3A |
| N.º de referencia del gen | P31941 |
| Alias de gen | A3A; Apobec3; APOBEC3A; apolipoprotein B editing complex 3; apolipoprotein B mRNA editing enzyme catalytic polypeptide like 3A; apolipoprotein B mRNA editing enzyme catalytic subunit 3A; apolipoprotein B mRNA editing enzyme, catalytic polypeptide 3; apolipoprotein B mRNA editing enzyme, catalytic polypeptide-like 3A; apolipoprotein B mRNA-editing complex 3; Arp3; BC003314; bK150C2.1; CEM15; CEM-15; DNA dC->dU-editing enzyme APOBEC-3; DNA dC->dU-editing enzyme APOBEC-3A; pho1; PHO-1; phorbolin 1; Phorbolin1; phorbolin-1; PHRBN; probable DNA dC->dU-editing enzyme APOBEC-3A; recovery from Friend virus 3; Rfv3 |
| Símbolos de los genes | APOBEC3A |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?