Learn More
DiscoverX Apelin-13 TFA salt
Código de producto. 17342429
Encompass
Change view
Click to view available options
Cantidad:
1 mg
Tamaño de la unidad:
1 miligramo
17342429 1 mg 1 miligramo
1 options
Código de producto. 17342429
Marca: DiscoverX 921093
Apelin-13 TFA salt is an agonist for AGTRL1. Provided as 1mg of dry powder alongside reconstitution buffer.
Apelin-13 is a bioactive neuropeptide processed from the pre-proprotein apelin that is active in the nervous system. It is an endogenous ligand of the receptor APJ. Plays a role in cardiovascular regulation, fluid homeostasis, hypertension, and insulin sensitivity. May undergo pyroglutamylation, and may block HIV-1 and HIV-2 entry into cells expressing the APJ receptor. Compared to apelin-36, blocks HIV entry poorly. Provided as 1mg of dry powder alongside reconstitution buffer.
Especificaciones
Tipo de producto Control Ligands