missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APBA1 Polyclonal Antibody

APBA1 Polyclonal Antibody
Marca: Invitrogen PA518702
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
APBA1 Polyclonal Antibody for Western Blot
Especificaciones
| APBA1 | |
| Polyclonal | |
| Unconjugated | |
| APBA1 | |
| 6430513E09Rik; adapter protein X11alpha; adaptor protein X11alpha; amyloid beta (A4) precursor protein binding, family A, member 1; amyloid beta (A4) precursor protein-binding family A APBA1: amyloid beta (A4) precursor protein-binding family A member 1 (X11); amyloid beta (A4) precursor protein-binding, family A, APBA1: amyloid beta (A4) precursor protein-binding, family A, member 1 (X11); amyloid beta (A4) precursor protein-binding, family A, member 1; amyloid beta (A4) precursor protein-binding, family A, member 1 (X11); amyloid beta A4 precursor protein-binding family A member 1; amyloid beta precursor protein binding family A member 1; Amyloid-beta A4 precursor protein-binding family A member 1; APBA1; D9S411E; LIN10; Lin-10; Mint; MINT1; Mint-1; neuronal Munc18-1-interacting protein 1; Neuron-specific X11 protein; phosphotyrosine-binding/-interacting domain (PTB)-bearing protein; Urop11; UROP11-110; X11; X11A; X11ALPHA | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 320 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q02410 | |
| APBA1 | |
| Synthetic peptide sequence (EASHPSQDGKRQYK) corresponding to the internal amino acids of APBA1 (aa 573-587). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido