Learn More
AP2B1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 15690-1-AP-150UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Subunit of clathrin-associated adaptor protein complex 1 that plays a role in protein sorting in the late-Golgi/trans-Golgi network (TGN) and/or endosomes. The AP complexes mediate both the recruitment of clathrin to membranes and the recognition of sorting signals within the cytosolic tails of transmembrane cargo molecules.Especificaciones
| AP2B1 | |
| Polyclonal | |
| Unconjugated | |
| AP2B1 | |
| ADTB2, AP 2 complex subunit beta, AP105B, AP2 BETA, AP2B1, Beta 2 adaptin, Beta adaptin, CLAPB1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 140670, 163, 71770 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P62944, P63010, Q9DBG3 | |
| Ap2b1 | |
| AP2B1 Fusion Protein Ag8248 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |