missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
Immunogen sequence: MYHALLKHLG TLAGSQIRNM ASLGGHIISR HPDSDLNPIL AVGNCTLNLL SKEGKRQIPL NEQFLSKCPN ADLKPQEILV SVNIPYSRKW E Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 86%.
AOX1 (Aldehyde oxidase 1) is a 1338 amino acid cytoplasmic protein that catalyzes the oxidation of a variety of aldehydes, leading to the production of hydrogen peroxide. Under certain conditions, AOX1 can catalyze the formation of the superoxide free radical. Defects in oxygen radical metabolism have been linked to the pathogenesis of amyotrophic lateral sclerosis (ALS), an autosomal dominant neurodegenerative disorder characterized by the death of motor neurons in the spinal cord, brain and brainstem. Significantly, AOX1 is highly expressed in the ventral horn of the spinal cord and the gene that encodes AOX1 is located in a chromosomal region that is frequently found to be implicated in ALS2. This evidence suggests that AOX1 is a candidate gene for ALS2.
Especificaciones
Especificaciones
| Antígeno | AOX1 |
| Aplicaciones | Immunohistochemistry (Paraffin) |
| Clasificación | Polyclonal |
| Concentración | 0.3 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| génica | AOX1 |
| N.º de referencia del gen | Q06278 |
| Alias de gen | AI196512; AI255253; Aldehyde oxidase; aldehyde oxidase (female form); aldehyde oxidase 1; aldehyde oxidase 2; Ao; AOH1; Aox1; Aox-1; Aox2; Aox-2; Azaheterocycle hydroxylase; azaheterocycle hydroxylase 1; Moro; Retinal oxidase; Ro |
| Símbolos de los genes | AOX1 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?
