Description
Removes the secondary (acyloxyacyl-linked) fatty acyl chains from the lipid A region of bacterial lipopolysaccharides.
Specifications
Specifications
| Antígeno | AOAH |
| Aplicaciones | Immunohistochemistry (Paraffin), Western Blot |
| Clasificación | Polyclonal |
| Concentración | 0.13 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 |
| génica | AOAH |
| N.º de referencia del gen | O35298, P28039 |
| Alias de gen | Acyloxyacyl hydrolase, AOAH |
| Símbolos de los genes | AOAH |
| Show More |