Learn More
ZAP-70 Kinase Mouse, Unlabeled, Clone: 29, BD
Descripción
ZAP-70 is a protein tyrosine kinase (PTK) that associates with the ζ subunit of the T cell antigen receptor (TCR) and undergoes tyrosine phosphorylation following TCR stimulation. ZAP-70 contains two SH2-like domains with the PTK domain being at the C-terminus. It appears that both ZAP-70 and Syk are recruited to the phosphorylated CD3 and ζ subunits after TCR stimulation. The src-family PTKs, lck, and fyn, seem to play a role upstream of ZAP-70 and Syk where they phosphorylate CD3 and ζ. Thus both Src- and ZAP-70/Syk-family PTKs are critical for efficient induction of tyrosine phosphoproteins in heterologous cells. The significance of ZAP-70 in mediating TCR signal transduction has been confirmed by showing that ZAP-70 activity is absent in an autosomal recessive form of severe combined immunodeficiency (SCID). This is due to mutations affecting the ZAP-70 kinase domain which affect the stability of the protein. The levels of lck and fyn appear to be normal in the same cells. Results of the ZAP-70 mutations in these patients include an unusual absence of peripheral CD8+ T cells and an abundance of CD4+ T cells in peripheral lymphoid organs. These results indicate that these two populations of T cells develop through utilization of different intracellular signaling pathways.
Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting
Especificaciones
Especificaciones
| Antígeno | ZAP-70 Kinase |
| Aplicaciones | Western Blot |
| Clasificación | Monoclonal |
| Clon | 29 |
| Concentración | 250μg/mL |
| Conjugado | Unconjugated |
| Formulación | Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. |
| Especie del huésped | Mouse |
| Inmunógeno | Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. |
| Método de purificación | Affinity Purified |
| Mostrar más |
For Research Use Only.
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.