Learn More
Vimentin Mouse anti-Human, PE, Clone: RV202, BD
Descripción
Intermediate filaments (IF) are a subset of cytoskeletal proteins which function to give overall structural integrity to the plasma membrane as well to organize cells into specific tissues. IF proteins can be divided into six major types based upon the similarity in sequence. Vimentin belongs to the type III category of IF proteins which are expressed in leukocytes, blood vessel endothelial cells, some epithelial cells, and mesenchymal cells. Vimentin is also expressed together with several other IF proteins during the early stages of development. Vimentin is exchanged for the tissue-specific intermediate filament type as differentiation proceeds. Vimentin migrates in SDS/PAGE as a ∽57 kDa protein.
Especificaciones
Especificaciones
| Antígeno | Vimentin |
| Aplicaciones | Flow Cytometry |
| Clasificación | Monoclonal |
| Clon | RV202 |
| Conjugado | PE |
| Formulación | Aqueous buffered solution containing BSA and ≤0.09% sodium azide. |
| Especie del huésped | Mouse |
| Inmunógeno | Purified Cow Lens Vimentin |
| Método de purificación | Affinity Purified |
| Cantidad | 50 Tests |
| Mostrar más |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.