Learn More
Vimentin Mouse, Unlabeled, Clone: RV202, BD
Mouse Monoclonal Antibody
Marca:  BD Biosciences 550513
Código de producto. 15826298
  • 279.00€ / cada uno
Descripción
Intermediate filaments (IF) are a subset of cytoskeletal proteins which function to give overall structural integrity to the plasma membrane as well to organize cells into specific tissues. IF proteins can be divided into six major types based upon the similarity in sequence. Vimentin belongs to the type III category of IF proteins which are expressed in leukocytes, blood vessel endothelial cells, some epithelial cells, and mesenchymal cells. Vimentin is also expressed together with several other IF proteins during the early stages of development. Vimentin is
exchanged for the tissue-specific intermediate filament type as differentiation proceeds. Vimentin migrates in SDS/PAGE as a ∽57kDa protein.
Host Species: Mouse
Clone: RV202
Isotype: IgG1
Species Reactivity: Human
Immunogen: Purified Cow Lens Vimentin
Electron Microscopy, Immunofluorescence, Immunohistochemistry, Western Blotting
TRUSTED_SUSTAINABILITY
Especificaciones
Especificaciones
Vimentin
Monoclonal
0.5mg/mL
Aqueous buffered solution containing ≤0.09% sodium azide.
Purified Cow Lens Vimentin
50 μg
Cell Biology
Bovine, Canine, Chicken, Goat, Hamster, Human, Monkey, Mouse, Rabbit, Rat
IgG1
Western Blot
RV202
Unconjugated
Mouse
Affinity Purified
RUO
Primary
Store undiluted at 4°C
Certificados
Promociones
Promociones
Título del producto
Seleccione un problema
For Research Use Only.