missing translation for 'onlineSavingsMsg'
Learn More
Learn More
transient receptor potential cation channel, subfamily C, member 3, Mouse, Polyclonal Antibody, Abnova™
Description
Sequence: TARFLAFLQATKAQQYVDSYVQESDLSEVTLPPEIQYFTYARDKWLPSDPQ
Specifications
Specifications
| Antígeno | transient receptor potential cation channel, subfamily C, member 3 |
| Aplicaciones | ELISA, Western Blot |
| Clasificación | Polyclonal |
| Conjugado | Unconjugated |
| Descripción | Mouse polyclonal antibody raised against a partial recombinant TRPC3. |
| Formulación | 50% glycerol |
| génica | TRPC3 |
| N.º de referencia del gen | NM_003305 |
| Alias de gen | TRP3 |
| Símbolos de los genes | TRPC3 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?