Learn More
Topo IIα Mouse anti-Human, Unlabeled, Clone: 31, BD
Mouse Monoclonal Antibody
451.00€
Especificaciones
| Antígeno | Topo IIα |
|---|---|
| Clon | 31 |
| Concentración | 250μg/mL |
| Aplicaciones | In vitro Assay, Western Blot |
| Clasificación | Monoclonal |
| Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
|---|---|---|---|---|---|---|---|---|---|
| Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
|
15865879
|
BD Biosciences
611327 |
150 μg |
451.00€
1 set |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | |||||
Descripción
Eukaryotic DNA topoisomerase II, a ubiquitous ATP-dependent type II topoisomerase, is an essential nuclear enzyme in DNA replication and transcription, chromatin segregation, and cell cycle progression. These enzymes transiently break a pair of complementary strands in double-stranded DNA to form a gate for the passage of duplex DNA. Two isoforms of DNA topoisomerase II have been identified, topo IIα and topo IIβ. These have a high degree of homology, except for some divergence in the C-terminal region. Both contain multiple bipartite nuclear localization sequences (NLS) that mediate their subnuclear localization. Topo IIα levels rise during late S phase and peak in G2-M, whereas topo IIβ levels remain constant throughout the cell cycle. In addition, topo IIα is expressed in proliferating cells, while topo IIβ is expressed in many tissues. The exact role of these two isoforms during cell proliferation is not known, however the differences in cellular expression implicate different physiological roles. Both isoforms may also be important targets for anticancer agents that exert cytotoxicity in proliferating cells via stabilization of a topo II-DNA complex.
Host Species: Mouse
Clone: 31
Isotype: IgG1
Species Reactivity: Human
Immunogen: Human Topo IIα aa. 1245-1361
Formula Weight [Chemical]: 170kDa
Immunofluorescence, Western Blotting
Especificaciones
| Topo IIα | |
| 250μg/mL | |
| Monoclonal | |
| Mouse | |
| Cell Biology | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| IgG1 | |
| Affinity Purified |
| 31 | |
| In vitro Assay, Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| Human Topo IIα aa. 1245-1361 | |
| Primary | |
| Store undiluted at -20°C. |
For Research Use Only.
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.