missing translation for 'onlineSavingsMsg'
Learn More

TNIK Mouse, Unlabeled, Clone: 53, BD

Código de producto. 15876139
Change view
Click to view available options
Cantidad:
50 μg
Tamaño de la unidad:
1 set
Código de producto. Cantidad unitSize
15876139 50 μg 1 set
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 15876139

Marca: BD Biosciences 612250

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Mouse Monoclonal Antibody

Ste20 is a S. cerevisiae Ser/Thr protein kinase that functions upstream of the MAP kinase module. Mammalian and yeast homologs of this kinase are divided into two classes based on their structure and regulation. Members of the first class (Ste20, Cla4, and p21-activated protein kinase) contain a C-terminal kinase domain, an N-terminal regulatory domain and a small GTPase Rac1/Cdc42 binding domain. Members of the second class lack the GTPase binding sites, but are similar to the former class throughout the catalytic domain. This second class includes germinal center kinase (GCK), HPK, KHS, KRS1 and 2, MST1, 2, and 3, NIK, SOK-1, and TNIK. Traf2- and NCK-interacting kinase (TNIK) is most homologous to another NCK-interacting kinase, NIK. TNIK contains an N- terminal kinase domain, and a C-terminal GCK homology (GCKH) domain. The mRNA of TNIK is expressed highest in heart, brain, and skeletal muscle. Overexpression of TNIK activates the JNK pathway, and leads to the disruption of F-actin structures and the inhibition of cell spreading. In vitro, TNIK can phosphorylate gelsolin. Thus, TNIK is a GCK family kinase that may regulate both the JNK pathway and cytoskeletal dynamics.

Western Blotting

TRUSTED_SUSTAINABILITY

Specifica

Antígeno TNIK
Aplicaciones Western Blot
Clasificación Monoclonal
Clon 53
Concentración 250μg/mL
Conjugado Unconjugated
Formulación Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Especie del huésped Mouse
Inmunógeno Human TNIK aa. 522-644
Método de purificación Affinity Purified
Cantidad 50 μg
Estado normativo RUO
Disciplina de investigación Cell Biology
Primario o secundario Primary
Especies diana Human, Mouse, Rat
Contenido y almacenamiento Store undiluted at -20°C.
Isotype IgG1
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.