missing translation for 'onlineSavingsMsg'
Learn More

TNF Mouse anti-Human, NA/LE, Unlabeled, Clone: MAB1, BD

Product Code. 10324640
Change view
Click to view available options
Cantidad:
0.5 mg
Unit Size:
cada uno
Product Code. Cantidad unitSize
10324640 0.5 mg cada uno
1 options
This item is not returnable. View return policy

Product Code. 10324640

Brand: BD Biosciences 554508

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Mouse Monoclonal Antibody

The MAb1 antibody reacts with human tumor necrosis factor (TNF, aka TNF-α) protein. The immunogen used to generate the MAb1 hybridoma was recombinant human TNF. The use of the MAb1 antibody for epitope mapping of human TNF has been described.

TRUSTED_SUSTAINABILITY

Specifications

Antígeno TNF
Aplicaciones ELISA
Clasificación Monoclonal
Clon MAB1
Concentración 1.0mg/mL
Conjugado Unconjugated
Formulación No azide/low endotoxin: Aqueous buffered solution containing no preservative, 0.2μm sterile filtered. Endotoxin level is ≤0.01 EU/μg (≤0.001 ng/μg) of protein as determined by the LAL assay.
Especie del huésped Mouse
Inmunógeno Recombinant Human TNF
Método de purificación Affinity Purified
Cantidad 0.5 mg
Estado normativo RUO
Disciplina de investigación Immunology
Primario o secundario Primary
Especies diana Human
Contenido y almacenamiento Store undiluted at 4°C
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.