Learn More
transcobalamin I (vitamin B12 binding protein, R binder family), Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human TCN1 protein.
Marca: Abnova H00006947-D01P.100ug
Description
This gene encodes a member of the vitamin B12-binding protein family. This family of proteins, alternatively referred to as R binders, is expressed in various tissues and secretions. This protein is a major constituent of secondary granules in neutrophils and facilitates the transport of cobalamin into cells. [provided by RefSeq
Sequence: MRQSHQLPLVGLLLFSFIPSQLCEICEVSEENYIRLKPLLNTMIQSNYNRGTSAVNVVLSLKLVGIQIQTLMQKMIQQIKYNVKSRLSDVSSGELALIILALGVCRNAEENLIYDYHLIDKLENKFQAEIENMEAHNGTPLTNYYQLSLDVLALCLFNGNYSTAEVVNHFTPENKNYYFGSQFSVDTGAMAVLALTCVKKSLINGQIKADEGSLKNISIYTKSLVEKILSEKKENGLIGNTFSTGEAMQALFVSSDYYNENDWNCQQTLNTVLTEISQGAFSNPNAAAQVLPALMGKTFLDINKDSSCVSASGNFNISADEPITVTPPDSQSYISVNYSVRINETYFTNVTVLNGSVFLSVMEKAQKMNDTIFGFTMEERSWGPYITCIQGLCANNNDRTYWELLSGGEPLSQGAGSYVVRNGENLEVRWSKYSpécification
| transcobalamin I (vitamin B12 binding protein, R binder family) | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against a full-length human TCN1 protein. | |
| TCN1 | |
| TC1/TCI | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 6947 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_001062.2 | |
| TCN1 | |
| TCN1 (NP_001053.2, 1 a.a. ∼ 433 a.a) full-length human protein. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.